missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trichlorobenzene, 99%, pure

Product Code. p-3737616 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10754641

Brand: Thermo Scientific Acros 157900010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 120-82-1 | C6H3Cl3 | 181.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-82-1
MDL Number MFCD00000547
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Melting Point 16.0°C
CAS Min % 98.5
CAS Max % 100.0
Density 1.4500g/mL
Boiling Point 214.0°C
Quantity 1 L
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.5700 to 1.5725
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.45
Molecular Formula C6H3Cl3
Beilstein 05, 204
Merck Index 15, 9796
Solubility Information Solubility in water: 49mg/l (20°C). Other solubilities: volatile with steam,sparingly soluble in alcohol,miscible with ether,benzene,petroleum ether and,carbon disulfide
Molecular Weight (g/mol) 181.45
Formula Weight 181.45
Percent Purity 99%
Grade Pure
Chemical Name or Material 1, 2, 4-Trichlorobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2,4-Trichlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.