missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trichlorobenzene, 99%, Thermo Scientific Chemicals

£26.60 - £229.00

Chemical Identifiers

CAS 120-82-1
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000547
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11469382
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10130.36
500 g
£26.60
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11479382
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10130.0E
2500 g
£77.50
2500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15473017
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10130.:L
4 x 2.5 kg
£229.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.

Solubility
Miscible with ether, benzene, CS{2}.Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

120-82-1
181.44
PBKONEOXTCPAFI-UHFFFAOYSA-N
13
1,2,4-trichlorobenzene
C6H3Cl3
MFCD00000547
benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
CHEBI:28222
C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

17°C
1.454
Liquid
C6H3Cl3
UN2321
benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PBKONEOXTCPAFI-UHFFFAOYSA-N
1,2,4-trichlorobenzene
13
181.45
1,2,4-Trichlorobenzene
120-82-1
213°C
1.572
MFCD00000547
14,9631
Miscible with ether,benzene,CS2;Immiscible with water.
C1=CC(=C(C=C1Cl)Cl)Cl
181.44
CHEBI:28222
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315
Harmful if swallowed.
Causes skin irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROBENZENES, LIQUID

missing translation for 'einecsNumber' : 204-428-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only