missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trichloro-5-iodobenzene, TRC

Product Code. 30457185
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457185 1 g 1g
30452556 10 g 10g
30450439 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457185 Supplier Toronto Research Chemicals Supplier No. TRCT7744251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7145-82-6
Molecular Formula C6H2Cl3I
Molecular Weight (g/mol) 307.34
Synonym 2,4,5-Trichloro-1-iodobenzene, 2,4,5-Trichloroiodobenzene, NSC 74997
IUPAC Name 1,2,4-trichloro-5-iodobenzene
SMILES Clc1cc(Cl)c(I)cc1Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H2Cl3I/c7-3-1-5(9)6(10)2-4(3)8/h1-2H
Formula Weight 305.8267
Chemical Name or Material 1,2,4-Trichloro-5-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.