missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Trichloro-5-iodobenzene, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Unit Size:
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1,2,4-Trichloro-5-iodobenzene can be used to produce 3-(2,4,5-trichloro-phenyl)-acrylic acid tert-butyl ester at the temperature of 80°C.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in the dark. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
1,2,4-Trichloro-5-iodobenzene can be used to produce 3-(2,4,5-trichloro-phenyl)-acrylic acid tert-butyl ester at the temperature of 80°C.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in the dark. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications
Specifications
| Melting Point | 102°C to 105°C |
| Boiling Point | 294°C (110 mmHg) |
| Quantity | 5 g |
| Beilstein | 1940881 |
| Sensitivity | Light sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 307.35 |
| Percent Purity | 98% |
| Chemical Name or Material | 1,2,4-Trichloro-5-iodobenzene |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu