missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Tributyl Phosphorotrithioate, TRC

Product Code. 30456056
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456056 50 mg 50mg
30457355 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456056 Supplier Toronto Research Chemicals Supplier No. TRCT88659350MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-48-8
Molecular Formula C12 H27 O P S3
Molecular Weight (g/mol) 314.51
Synonym Butyl phosphorotrithioate (6CI), B 1,776, Butiphos, Butyl phosphorotrithioate ((BuS)3PO), Chemagro B 1776, DEF, DEF (defoliant), DEF 6, De-Green, Fos-Fall A, Fosfall, S,S,S-Tributyl phosphorotrithioate, S,S,S-Tributyl trithiophosphate, TBPT, TBTP, Tribufos
IUPAC Name 1-bis(butylsulfanyl)phosphorylsulfanylbutane
SMILES CCCCSP(=O)(SCCCC)SCCCC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H27OPS3/c1-4-7-10-15-14(13,16-11-8-5-2)17-12-9-6-3/h4-12H2,1-3H3
Formula Weight 314.0962
Chemical Name or Material Tribufos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.