missing translation for 'onlineSavingsMsg'
Learn More

[1,2,4]Triazolo[1,5-a]pyridine-6-carboxylic Acid, TRC

Product Code. 30455723
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455723 100 mg -
30453995 10 mg 10mg
30451990 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455723 Supplier Toronto Research Chemicals Supplier No. TRCT795823100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1043903-19-0
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.13
IUPAC Name [1,2,4]triazolo[1,5-a]pyridine-6-carboxylic acid
SMILES OC(=O)c1ccc2ncnn2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H5N3O2/c11-7(12)5-1-2-6-8-4-9-10(6)3-5/h1-4H,(H,11,12)
Formula Weight 163.0382
Chemical Name or Material [1,2,4]Triazolo[1,5-a]pyridine-6-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.