missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazole, sodium salt, 90%, technical

Product Code. 10173820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173820 50 g Glass bottle 50g
10341004 10 g Glass bottle 10g
10091881 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10173820 Supplier Thermo Scientific Acros Supplier No. 188990500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41253-21-8 | C2H2N3Na | 91.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41253-21-8
Molecular Formula C2H2N3Na
Molecular Weight (g/mol) 91.05
MDL Number MFCD00044437
InChI Key NVMNEWNGLGACBB-UHFFFAOYSA-N
Synonym sodium 1,2,4-triazol-1-ide, 1,2,4-triazolylsodium, 1,2,4-triazole sodium derivative, 1,2,4-triazole sodium salt, sodium 1,2,4-triazolide, 1h-1,2,4-triazole, sodium salt, sodium 1h-1,2,4-triazolate, 1,2,4-triazole sodium, sodium 1h-1,2,4-triazolide, 1,2,4-triazole, sodium salt
PubChem CID 4649906
IUPAC Name sodium;2,4-diaza-1-azanidacyclopenta-2,4-diene
SMILES C1=NC=N[N-]1.[Na+]

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 295°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 50 g
Beilstein 26, II, 7
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in most organic solvents
Formula Weight 91.05
Percent Purity 90%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4-Triazole, sodium derivative, tech., 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2,4-Triazole, sodium derivative
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.