missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Triazole-d2, TRC
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Specifications
Specifications
| Chemical Name or Material | 1,2,4-Triazole-d2 |
| Molecular Formula | C2 D2 H N3 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C2H3N3/c1-3-2-5-4-1/h1-2H,(H,3,4,5)/i1D,2D |
| SMILES | [2H]c1n[nH]c([2H])n1 |
| IUPAC Name | 3,5-dideuterio-1H-1,2,4-triazole |
| Molecular Weight (g/mol) | 71.078 |
| Formula Weight | 71.0453 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction