missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triaminobenzene Dihydrochloride, TRC

Product Code. 30455962
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455962 500 mg 500mg
30449079 100 mg 100mg
30454244 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455962 Supplier Toronto Research Chemicals Supplier No. TRCT895865500MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-47-4
Molecular Formula C6H9N3 . 2(HCl)
Molecular Weight (g/mol) 196.07
Synonym 1,2,4-Triamino-Benzene 2HCl
IUPAC Name benzene-1,2,4-triamine;dihydrochloride
SMILES Cl.Cl.Nc1ccc(N)c(N)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H9N3.2ClH/c7-4-1-2-5(8)6(9)3-4;;/h1-3H,7-9H2;2*1H
Formula Weight 195.033
Chemical Name or Material 1,2,4-Triaminobenzene Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.