missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Benzenetriol, TRC

Product Code. 30007023
Change view
Click to view available options
Quantity:
10 mg
1 g
5 g
Unit Size:
10mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007023 10 mg 10mg
30006442 1 g 1g
30005567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007023 Supplier Toronto Research Chemicals Supplier No. TRCB18980010MG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-73-3
Molecular Formula C6 H6 O3
Molecular Weight (g/mol) 126.11
Synonym 1,2,4-Benzenetriol, 1,2,4-Trihydroxybenzene, 1,3,4-Benzenetriol, 1,3,4-Trihydroxybenzene, 2,5-Dihydroxyphenol, 2-Hydroxy-1,4-hydroquinone, 2-Hydroxy-p-benzohydroquinone, 2-Hydroxyhydroquinone, 4-Hydroxycatechol, HHQ, Hydroxyhydroquinone, NSC 2818
IUPAC Name benzene-1,2,4-triol
SMILES Oc1ccc(O)c(O)c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C6H6O3/c7-4-1-2-5(8)6(9)3-4/h1-3,7-9H
Formula Weight 126.0317
Chemical Name or Material 1,2,4-Benzenetriol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.