missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Benzenetricarboxylic Acid 1-(2-ethylhexyl) Ester, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 1,2,4-Benzenetricarboxylic Acid 1-(2-ethylhexyl) Ester |
| CAS | 61137-09-5 |
| Molecular Formula | C17 H22 O6 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C17H22O6/c1-3-5-6-11(4-2)10-23-17(22)13-8-7-12(15(18)19)9-14(13)16(20)21/h7-9,11H,3-6,10H2,1-2H3,(H,18,19)(H,20,21) |
| SMILES | CCCCC(CC)COC(=O)c1ccc(cc1C(=O)O)C(=O)O |
| IUPAC Name | 4-(2-ethylhexoxycarbonyl)benzene-1,3-dicarboxylic acid |
| Molecular Weight (g/mol) | 322.353 |
| Formula Weight | 322.142 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction