missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Benzenetricarboxylic Acid 1,2-Bis(2-ethylhexyl) Ester, TRC

Product Code. 30138183
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138183 100 mg 100mg
30137980 2.5 mg 2.5mg
30012580 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138183 Supplier Toronto Research Chemicals Supplier No. TRCB190030100MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63468-09-7
Molecular Formula C25 H38 O6
Molecular Weight (g/mol) 434.57
IUPAC Name 3,4-bis(2-ethylhexoxycarbonyl)benzoic acid
SMILES CCCCC(CC)COC(=O)c1ccc(cc1C(=O)OCC(CC)CCCC)C(=O)O

Specifications

InChI Formula InChI=1S/C25H38O6/c1-5-9-11-18(7-3)16-30-24(28)21-14-13-20(23(26)27)15-22(21)25(29)31-17-19(8-4)12-10-6-2/h13-15,18-19H,5-12,16-17H2,1-4H3,(H,26,27)
Formula Weight 434.2668
Quantity 100 mg
Chemical Name or Material 1,2,4-Benzenetricarboxylic Acid 1,2-Bis(2-ethylhexyl) Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.