missing translation for 'onlineSavingsMsg'
Learn More

1-(2,4,6-Triisopropylphenylsulfonyl)-1,2,4-triazole, 98%, Thermo Scientific™

Product Code. 11476007
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11476007

Brand: Thermo Scientific Alfa Aesar B25038.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54230-60-3
Molecular Formula C17H25N3O2S
Molecular Weight (g/mol) 335.47
MDL Number MFCD00014089
InChI Key BKJMMUUBRUWPPQ-UHFFFAOYSA-N
Synonym 1-2,4,6-tris isopropyl phenyl sulphonyl-1h-1,2,4-triazole, 1-2,4,6-triisopropylphenyl sulfonyl-1h-1,2,4-triazole, 1-2,4,6-triisopropylbenzenesulfonyl-1,2,4-triazole, tpst, 2,4,6-triisopropylbenzenesulfonyl-1h-1,2,4-triazole, 1-2,4,6-triisopropylphenyl-sulfonyl-1h-1,2,4-triazole, 1-2,4,6-triisopropylphenylsulfonyl-1,2,4-triazole, 1-2,4,6-tri propan-2-yl phenyl sulfonyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-2,4,6-tris 1-methylethyl phenyl sulfonyl
PubChem CID 688654
IUPAC Name 1-[2,4,6-tri(propan-2-yl)phenyl]sulfonyl-1,2,4-triazole
SMILES CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(=O)(=O)N1C=NC=N1

Specifications

Melting Point 110°C to 112°C
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 335.47
Percent Purity 98%
Chemical Name or Material 1-(2,4,6-Triisopropylphenylsulfonyl)-1,2,4-triazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.