missing translation for 'onlineSavingsMsg'
Learn More

1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole, 98%, Thermo Scientific™

Product Code. 15425796
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15425796

Brand: Thermo Scientific Alfa Aesar H60482.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50257-40-4
Molecular Formula C18H26N2O2S
Molecular Weight (g/mol) 334.478
MDL Number MFCD00005283
InChI Key AGGRGODMKWLSDE-UHFFFAOYSA-N
Synonym 1-2,4,6-triisopropylphenyl sulfonyl-1h-imidazole, 1-2,4,6-triisopropylbenzenesulfonyl imidazole, 1-2,4,6-triisopropylphenylsulfonyl imidazole, tpsi, 1-2',4',6'-triisopropylbenzenesulphonyl imidazole, 1-2,4,6-tri propan-2-yl phenyl sulfonylimidazole, tpsi 1-2,4,6-triisopropylbenzenesulfonyl imidazole, 1h-imidazole, 1-2,4,6-tris 1-methylethyl phenyl sulfonyl, 1-2,4,6-trisopropylbenzenesulfonyl imidazole
PubChem CID 521274
IUPAC Name 1-[2,4,6-tri(propan-2-yl)phenyl]sulfonylimidazole
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N2C=CN=C2)C(C)C

Specifications

Melting Point 118°C to 122°C
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 334.48
Percent Purity 98%
Chemical Name or Material 1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.