missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrafluorobenzene, 99%

Product Code. 11473408
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473408 5 g 5g
11483408 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473408 Supplier Thermo Scientific Alfa Aesar Supplier No. A17815.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 327-54-8 | C6H2F4 | 150.076 g/mol

It is employed as intermediate in organic synthesis. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate in organic synthesis. It is also used as pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327-54-8
Molecular Formula C6H2F4
Molecular Weight (g/mol) 150.076
MDL Number MFCD00000307
InChI Key SDXUIOOHCIQXRP-UHFFFAOYSA-N
Synonym benzene, 1,2,4,5-tetrafluoro, 2,3,5,6-tetrafluorobenzene, pubchem1063, acmc-209hvc, 1,4,5-tetrafluorobenzene, 2,5,6-tetrafluorobenzene, 2,3,5,6-tetrafluorophenyl, ksc496i2b, benzene,2,4,5-tetrafluoro, 1,2,4,5-tetrafluoro-benzene
PubChem CID 9474
IUPAC Name 1,2,4,5-tetrafluorobenzene
SMILES C1=C(C(=CC(=C1F)F)F)F

Specifications

Melting Point 3°C to 5°C
Density 1.43
Boiling Point 90°C
Flash Point 16°C (60°F)
Refractive Index 1.407
Quantity 5 g
UN Number UN2387
Beilstein 1909052
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 150.07
Percent Purity 99%
Chemical Name or Material 1,2,4,5-Tetrafluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.