missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrachlorobenzene, TRC

Product Code. 30180246
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180246 10 g 10g
30179928 50 g 50g
30179735 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180246 Supplier Toronto Research Chemicals Supplier No. TRCT29128510G
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-94-3
Molecular Formula C6 H2 Cl4
Molecular Weight (g/mol) 215.89
Synonym Benzene, 1,2,4,5-tetrachloro-, 1,2,4,5-Tetrachlorobenzene, NSC 27003, s-Tetrachlorobenzene
IUPAC Name 1,2,4,5-tetrachlorobenzene
SMILES Clc1cc(Cl)c(Cl)cc1Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H2Cl4/c7-3-1-4(8)6(10)2-5(3)9/h1-2H
Formula Weight 213.8911
Chemical Name or Material 1,2,4,5-Tetrachlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.