missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrachlorobenzene, 98%

Product Code. 11426867
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. Quantity unitSize
11426867 250 g 250g
11484240 10 g 10g
11494240 50 g 50g
3 options
This item is not returnable. View return policy

Product Code. 11426867

Brand: Thermo Scientific Alfa Aesar A14318.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 95-94-3 | C6H2Cl4 | 215.882 g/mol

It finds its application as a high melting diluent in the eutectic crystallization of pseudo binary systems of polyethylene and high melting diluents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a high melting diluent in the eutectic crystallization of pseudo binary systems of polyethylene and high melting diluents.

Solubility
Insoluble in water. Solubility in toluene is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-94-3
Molecular Formula C6H2Cl4
Molecular Weight (g/mol) 215.882
MDL Number MFCD00000549
InChI Key JHBKHLUZVFWLAG-UHFFFAOYSA-N
Synonym s-tetrachlorobenzene, benzene, 1,2,4,5-tetrachloro, benzene tetrachloride, rcra waste number u207, 1,2,4,5-tetrachlorbenzol, unii-5n27529kgh, ccris 766, benzene, tetrachloro, rcra waste no. u207, 1,2,4,5-tetrachlorbenzol russian
PubChem CID 7270
ChEBI CHEBI:36697
IUPAC Name 1,2,4,5-tetrachlorobenzene
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)Cl

Specifications

Melting Point 138°C to 142°C
Density 1.86
Boiling Point 245°C
Quantity 250 g
Odor Unpleasant
UN Number UN3077
Beilstein 1618315
Solubility Information Insoluble in water. Solubility in toluene is almost transparent.
Formula Weight 215.89
Percent Purity 98%
Chemical Name or Material 1,2,4,5-Tetrachlorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.