missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals

Catalog Number 10213990
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
89-05-4
C10H6O8
254.15
MFCD00002471
CYIDZMCFTVVTJO-UHFFFAOYSA-N
pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene
6961
CHEBI:45165
benzene-1,2,4,5-tetracarboxylic acid
C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O
This item is not returnable. View return policy
89-05-4
C10H6O8
254.15
MFCD00002471
CYIDZMCFTVVTJO-UHFFFAOYSA-N
pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene
6961
CHEBI:45165
benzene-1,2,4,5-tetracarboxylic acid
C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 281.0°C to 284.5°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula C6H2(CO2H)4
Quantity 5 g
Beilstein 09, 997
Merck Index 15, 8116
Solubility Information Solubility in water: 1.5g/100mL water (20°C). Other solubilities: 30g/100mL water (100°C),freely soluble in alcohol
Formula Weight 254.15
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4,5-Benzenetetracarboxylic acid, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2,4,5-Benzenetetracarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'