missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Benzenetetracarboxylic acid, 96%

Product Code. 10040570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10040570 100 g Plastic bottle 100g
10213990 5 g Glass bottle 5g
10132370 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10040570 Supplier Thermo Scientific Acros Supplier No. 105311000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-05-4 | C10H6O8 | 254.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-05-4
Molecular Formula C10H6O8
Molecular Weight (g/mol) 254.15
MDL Number MFCD00002471
InChI Key CYIDZMCFTVVTJO-UHFFFAOYSA-N
Synonym pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene
PubChem CID 6961
ChEBI CHEBI:45165
IUPAC Name benzene-1,2,4,5-tetracarboxylic acid
SMILES C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O

Especificaciones

CAS Min % 95.0
CAS Max % 100.0
Melting Point 281.0°C to 284.5°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic bottle
Linear Formula C6H2(CO2H)4
Quantity 100 g
Beilstein 09, 997
Merck Index 15, 8116
Solubility Information Solubility in water: 1.5g/100mL water (20°C). Other solubilities: 30g/100mL water (100°C),freely soluble in alcohol
Formula Weight 254.15
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4,5-Benzenetetracarboxylic acid, 96%
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 1,2,4,5-Benzenetetracarboxylic acid
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.