missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trimethylimidazolium methyl sulfate, 95%, Thermo Scientific™

Product Code. p-7053361
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15496118

Brand: Thermo Scientific Alfa Aesar H59724.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65086-12-6
Molecular Formula C7H14N2O4S
Molecular Weight (g/mol) 222.259
MDL Number MFCD03788934
InChI Key OUAUEIYYLHUEPK-UHFFFAOYSA-M
Synonym 1,2,3-trimethylimidazolium methyl sulfate, basionic tm st 99, 1,2,3-trimethylimidazol-1-ium methyl sulfate 1-, basionics™ st 99, 1,2,3-trimethylimidazolium methanesulfonate, 1,2,3-trimethylimidazolium methyl sulfate, basf quality
PubChem CID 11579446
IUPAC Name methyl sulfate;1,2,3-trimethylimidazol-1-ium
SMILES CC1=[N+](C=CN1C)C.COS(=O)(=O)[O-]

Specifications

Melting Point 115°C
Flash Point 196°C (385°F)
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 222.26
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1,2,3-Trimethylimidazolium methyl sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.