missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trimethoxybenzene, TRC

Product Code. 30132678
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132678 50 g 50g
30003940 10 g 10g
30009981 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132678 Supplier Toronto Research Chemicals Supplier No. TRCT89600050G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634-36-6
Molecular Formula C9 H12 O3
Molecular Weight (g/mol) 168.19
Synonym Benzene, 1,2,3-trimethoxy- (6CI, 7CI, 8CI, 9CI, ACI), 1,2,3-Trimethoxybenzene (ACI), 1,2,6-Trimethoxybenzene, 3,4,5-Trimethoxybenzole, Methylsyringol, NSC 10124, Pyrogallol trimethoxy ether, Pyrogallol trimethyl ether, Tri-O-methylpyrogallol
IUPAC Name 1,2,3-trimethoxybenzene
SMILES COc1cccc(OC)c1OC

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H12O3/c1-10-7-5-4-6-8(11-2)9(7)12-3/h4-6H,1-3H3
Formula Weight 168.0786
Chemical Name or Material 1,2,3-Trimethoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.