missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trimethoxybenzene, 98%

Product Code. 10317770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10317770 25 g Glass bottle 25g
10605412 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10317770 Supplier Thermo Scientific Acros Supplier No. 160210250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 634-36-6 | C9H12O3 | 168.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634-36-6
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00008358
InChI Key CRUILBNAQILVHZ-UHFFFAOYSA-N
Synonym methylsyringol, pyrogallol trimethyl ether, trimethoxybenzene, benzene, 1,2,3-trimethoxy, tri-o-methylpyrogallol, benzene, trimethoxy, unii-mre1o894fg, 1,2,3-trimethoxy benzene, mre1o894fg, 3,4,5-trimethoxybenzene
PubChem CID 12462
ChEBI CHEBI:86529
IUPAC Name 1,2,3-trimethoxybenzene
SMILES COC1=C(C(=CC=C1)OC)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 42.0°C to 45.0°C
Color Beige to White
Density 1.1120g/mL
Boiling Point 241.0°C
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(OCH3)3
Quantity 25 g
Beilstein 06, 1081
Specific Gravity 1.112
Solubility Information Solubility in water: insoluble
Formula Weight 168.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 2, 3-Trimethoxybenzene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.