missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trimethoxybenzene, 98+%

Product Code. 11441968
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441968 500 g 500g
11421968 25 g 25g
11431968 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441968 Supplier Thermo Scientific Alfa Aesar Supplier No. A14572.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 634-36-6 | C9H12O3 | 168.192 g/mol

1,2,3-Trimethoxybenzene was used to study the effect of solvent on photo induced electron-transfer reactions, on condensation with 2,4-diamino-5-(hydroxymethyl)pyrimidine yields 2,4-diamino-5-(2,3,4-trimethoxybenzyl)pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3-Trimethoxybenzene was used to study the effect of solvent on photo induced electron-transfer reactions, on condensation with 2,4-diamino-5-(hydroxymethyl)pyrimidine yields 2,4-diamino-5-(2,3,4-trimethoxybenzyl)pyrimidine.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634-36-6
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00008358
InChI Key CRUILBNAQILVHZ-UHFFFAOYSA-N
Synonym methylsyringol, pyrogallol trimethyl ether, trimethoxybenzene, benzene, 1,2,3-trimethoxy, tri-o-methylpyrogallol, benzene, trimethoxy, unii-mre1o894fg, 1,2,3-trimethoxy benzene, mre1o894fg, 3,4,5-trimethoxybenzene
PubChem CID 12462
ChEBI CHEBI:86529
IUPAC Name 1,2,3-trimethoxybenzene
SMILES COC1=C(C(=CC=C1)OC)OC

Specifications

Melting Point 43°C to 45°C
Density 1.112
Boiling Point 241°C
Flash Point 112°C (233°F)
Quantity 500 g
Beilstein 1910422
Solubility Information Insoluble in water.
Formula Weight 168.19
Percent Purity ≥98%
Chemical Name or Material 1,2,3-Trimethoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.