missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trifluorobenzene, 98+%

Product Code. p-7026193
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11499547

Brand: Thermo Scientific Alfa Aesar B22055.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1489-53-8 | C6H3F3 | 132.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1489-53-8
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.085
MDL Number MFCD00012013
InChI Key AJKNNUJQFALRIK-UHFFFAOYSA-N
Synonym trifluorobenzene, benzene, trifluoro, benzene, 1,2,3-trifluoro, 3,4,5-trifluoro-benzene, pubchem1067, 1,2,3-fluorobenzene, 1,2,3-trifluorobenze, 1,2,3-trifluorbenzen, 1,2,6-trifluorobenzene, 2,3,4-trifluorobenzene
PubChem CID 15147
IUPAC Name 1,2,3-trifluorobenzene
SMILES C1=CC(=C(C(=C1)F)F)F

Specifications

Density 1.28
Boiling Point 94°C to 95°C
Flash Point −3°C (26°F)
Refractive Index 1.422
Quantity 1 g
UN Number UN2387
Beilstein 1862388
Formula Weight 132.09
Percent Purity ≥98%
Chemical Name or Material 1,2,3-Trifluorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.