missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3-Trichloropropane, 98+%
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
Description
1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.
Solubility
Slightly soluble in water (2 g/L at 25°C).
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.
Solubility
Slightly soluble in water (2 g/L at 25°C).
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
Specifications
Specifications
| Melting Point | -14°C |
| Density | 1.385 |
| Boiling Point | 156°C to 158°C |
| Flash Point | 74°C (165°F) |
| Refractive Index | 1.4822 |
| Quantity | 250 g |
| UN Number | UN2810 |
| Beilstein | 1732068 |
| Solubility Information | Slightly soluble in water (2g/L at 25°C). |
| Formula Weight | 147.43 |
| Show More |
Safety and Handling
Product Identifier
- 1,2,3-trichloropropane
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H350-May cause cancer.
- H360F-May damage fertility.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : 1,2,3-Trichloropropane
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction