missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3-Trichlorobenzene, TRC
Click to view available options
Quantity:
250 g
Unit Size:
250g
Specifications
Specifications
| Chemical Name or Material | 1,2,3-Trichlorobenzene |
| CAS | 87-61-6 |
| Molecular Formula | C6 H3 Cl3 |
| Quantity | 250 g |
| Synonym | Benzene','1,2,3-trichloro-, 1,2,3-Trichlorobenzene, 1,2,6-Trichlorobenzene, NSC 43432, vic-Trichlorobenzene |
| InChI Formula | InChI=1S/C6H3Cl3/c7-4-2-1-3-5(8)6(4)9/h1-3H |
| SMILES | Clc1cccc(Cl)c1Cl |
| IUPAC Name | 1,2,3-trichlorobenzene |
| Molecular Weight (g/mol) | 181.45 |
| Formula Weight | 179.93 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction