missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trichlorobenzene, 99%

Product Code. 11489677
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11489677

Brand: Thermo Scientific Alfa Aesar B22154.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 87-61-6 | C6H3Cl3 | 181.44 g/mol

Detoxification by catalytic hydrotreatment of 1,2,3-Trichlorobenzene is used for the disposal of hazardous organic waste liquids. As constituent of trichlorobenzene mixt used for termite control. As transformer fluid, dye carrier & solvent. Solvent for high melting products, Coolant in electrical installations and glass tempering. In polyester dyeing, lubricants, Heat transfer medium. As chemical intermediate for 2,3-dichlorophenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Detoxification by catalytic hydrotreatment of 1,2,3-Trichlorobenzene is used for the disposal of hazardous organic waste liquids. As constituent of trichlorobenzene mixt used for termite control. As transformer fluid, dye carrier & solvent. Solvent for high melting products, Coolant in electrical installations and glass tempering. In polyester dyeing, lubricants, Heat transfer medium. As chemical intermediate for 2,3-dichlorophenol.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-61-6
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000537
InChI Key RELMFMZEBKVZJC-UHFFFAOYSA-N
Synonym trichlorobenzene, vic-trichlorobenzene, 1,2,6-trichlorobenzene, benzene, 1,2,3-trichloro, benzene, trichloro, unii-nur9777ik4, ccris 5944, dsstox_cid_6193, dsstox_rid_78052, dsstox_gsid_26193
PubChem CID 6895
ChEBI CHEBI:35289
IUPAC Name 1,2,3-trichlorobenzene
SMILES ClC1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 53°C to 55°C
Density 1.69
Boiling Point 218°C to 219°C
Quantity 100 g
UN Number UN3077
Beilstein 956882
Merck Index 14,9630
Solubility Information Insoluble in water.
Formula Weight 181.45
Percent Purity 99%
Chemical Name or Material 1,2,3-Trichlorobenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.