missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trichlorobenzene 99%, Thermo Scientific™

Product Code. 10605002 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
5g
This item is not returnable. View return policy

Product Code. 10605002

Brand: Thermo Scientific Acros 139390050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-61-6
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000537
InChI Key RELMFMZEBKVZJC-UHFFFAOYSA-N
Synonym trichlorobenzene, vic-trichlorobenzene, 1,2,6-trichlorobenzene, benzene, 1,2,3-trichloro, benzene, trichloro, unii-nur9777ik4, ccris 5944, dsstox_cid_6193, dsstox_rid_78052, dsstox_gsid_26193
PubChem CID 6895
ChEBI CHEBI:35289
IUPAC Name 1,2,3-trichlorobenzene
SMILES ClC1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 50°C to 54°C
CAS Min % 98.5
Color White
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Assay Percent Range 99%
Beilstein 05, 203
Merck Index 15, 9795
Formula Weight 181.45
Percent Purity ≥98.5%
Chemical Name or Material 1, 2, 3-Trichlorobenzene, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.