Learn More
1,2,3-Thiadiazole-4-carbaldehyde, Thermo Scientific™
Brand: Thermo Scientific Maybridge SP01373CB
Code : 31
Additional Details : CAS Number : 27643-15-8 Weight : 0.05025kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
27643-15-8 | |
114.122 | |
1,2,3-thiadiazole-4-carbaldehyde, 1,2,3-thiadiazole-4-carboxaldehyde, 4-thiadiazolecarboxaldehyde, acmc-1cb1x, 1,2,3-thiadiazol-4-carboxaldehyde | |
thiadiazole-4-carbaldehyde |
C3H2N2OS | |
JNEBZFFTOLBIKJ-UHFFFAOYSA-N | |
2737271 | |
C1=C(N=NS1)C=O |
Specifications
1, 2, 3-Thiadiazole-4-carbaldehyde | |
97% | |
Amber Glass Bottle | |
250mg | |
C1=C(N=NS1)C=O | |
114.122 | |
114.13 |
27643-15-8 | |
C3H2N2OS | |
1,2,3-thiadiazole-4-carbaldehyde, 1,2,3-thiadiazole-4-carboxaldehyde, 4-thiadiazolecarboxaldehyde, acmc-1cb1x, 1,2,3-thiadiazol-4-carboxaldehyde | |
JNEBZFFTOLBIKJ-UHFFFAOYSA-N | |
thiadiazole-4-carbaldehyde | |
2737271 | |
97% |