missing translation for 'onlineSavingsMsg'
Learn More

1-[(2,3-Dihydro-1,4-benzodioxin-2-yl)carbonyl]piperazine Monohydrochloride, TRC

Product Code. 30386399
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386399 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386399 Supplier Toronto Research Chemicals Supplier No. TRCD4484451G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[(2RS)-2,3-Dihydro-1,4-benzodioxin-2-ylcarbonyl]piperazine Hydrochloride
CAS 70918-74-0
Molecular Formula C13 H16 N2 O3 . Cl H
Quantity 1 g
Synonym 1-[(2RS)-2,3-Dihydro-1,4-benzodioxin-2-ylcarbonyl]piperazine Hydrochloride, Doxazosin Mesilate Imp. B (EP) as Hydrochloride, Methanone, (2,3-dihydro-1,4-benzodioxin-2-yl)-1-piperazinyl-, hydrochloride (1:1), Piperazine, 1-[(2,3-dihydro-1,4-benzodioxin-2-yl)carbonyl]-, monohydrochloride (9CI), 1,4-Benzodioxin, piperazine deriv., 1-[(2,3-Dihydro-1,4-benzodioxin-2-yl)carbonyl]piperazine Monohydrochloride, 1-(1,4-Benzodioxan-2-ylcarbonyl)piperazine hydrochloride
InChI Formula InChI=1S/C13H16N2O3.ClH/c16-13(15-7-5-14-6-8-15)12-9-17-10-3-1-2-4-11(10)18-12;/h1-4,12,14H,5-9H2;1H
SMILES Cl.O=C(C1COc2ccccc2O1)N3CCNCC3
IUPAC Name 2,3-dihydro-1,4-benzodioxin-3-yl(piperazin-1-yl)methanone;hydrochloride
Molecular Weight (g/mol) 284.74
Formula Weight 284.0928
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.