missing translation for 'onlineSavingsMsg'
Learn More

1-(2,3-Dichlorophenyl)piperazine monohydrochloride, 98+%, Thermo Scientific™

Product Code. 11399465
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399465 100 g 100g
11389465 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399465 Supplier Thermo Scientific Alfa Aesar Supplier No. L18697.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
This product is intended for forensic and research purposes and it is an aripiprazole intermediate.

Solubility
Soluble in water, DMSO and methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119532-26-2
Molecular Formula C10H13Cl3N2
Molecular Weight (g/mol) 267.578
MDL Number MFCD00190238
InChI Key CYQFNNSFAGXCEC-UHFFFAOYSA-N
Synonym 1-2,3-dichlorophenyl piperazine hydrochloride, 1-2,3-dichlorophenyl piperazine monohydrochloride, 1-2,3-dichlorophenyl-piperazine hydrochloride, 2,3-dichlorophenylpiperazine hydrochloride, 1-2,3-dichlorophenyl-piperazine monohydrochloride, 1-2,3-dichlorophenyl piperazinehydrochloride, piperazine, 1-2,3-dichlorophenyl-, monohydrochloride, 1-2,3-dichlorophenyl piperazine;hydrochloride, 2,3-dichlorophenyl piperazine, chloride, zlchem 392
PubChem CID 11630372
IUPAC Name 1-(2,3-dichlorophenyl)piperazine;hydrochloride
SMILES C1CN(CCN1)C2=C(C(=CC=C2)Cl)Cl.Cl

Specifications

Melting Point 248°C to 250°C
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,DMSO and methanol.
Formula Weight 267.59
Percent Purity ≥98%
Chemical Name or Material 1-(2,3-Dichlorophenyl)piperazine monohydrochloride

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.