missing translation for 'onlineSavingsMsg'
Learn More

1-(2,3-Dichloro-4-methoxyphenyl)-1-butanone-d7, TRC

Product Code. 30377723
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377723 100 mg 100mg
30384339 25 mg 25mg
30381315 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30377723 Supplier Toronto Research Chemicals Supplier No. TRCD434972100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2,3-Dichloro-4-methoxyphenyl)-1-butanone-d7
Molecular Formula C11 D7 H5 Cl2 O2
Quantity 100 mg
InChI Formula InChI=1S/C11H12Cl2O2/c1-3-4-8(14)7-5-6-9(15-2)11(13)10(7)12/h5-6H,3-4H2,1-2H3/i1D3,3D2,4D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)c1ccc(OC)c(Cl)c1Cl
IUPAC Name 2,2,3,3,4,4,4-heptadeuterio-1-(2,3-dichloro-4-methoxyphenyl)butan-1-one
Molecular Weight (g/mol) 254.161
Formula Weight 253.065
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.