missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,7,8,9-Hexachlorodibenzofuran, TRC

Product Code. 30180338
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180338 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30180338 Supplier Toronto Research Chemicals Supplier No. TRCH2650905MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 72918-21-9
Molecular Formula C12 H2 Cl6 O
Quantity 5 mg
Synonym Dibenzofuran, 1,2,3,7,8,9-hexachloro-, 1,2,3,7,8,9-Hexachlorodibenzofuran, 1,2,3,7,8,9-HxCDF, F 124, PCDF 124, 1,2,3,7,8,9-HexaCDF, 123789-HexaCDF, 123789-HxCDF, 123789-Hexachlorodibenzofuran
InChI Formula InChI=1S/C12H2Cl6O/c13-3-1-5-7(11(17)9(3)15)8-6(19-5)2-4(14)10(16)12(8)18/h1-2H
SMILES Clc1cc2oc3cc(Cl)c(Cl)c(Cl)c3c2c(Cl)c1Cl
IUPAC Name 1,2,3,7,8,9-hexachlorodibenzofuran
Molecular Weight (g/mol) 374.86
Formula Weight 371.8237
Chemical Name or Material 1,2,3,7,8,9-Hexachlorodibenzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.