missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,6-Tetrahydropyridine, TRC

Product Code. 30452655
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452655 25 mg 25mg
30451028 50 mg 50mg
30451372 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30452655 Supplier Toronto Research Chemicals Supplier No. TRCT79621825MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-05-3
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
Synonym 1,2,3,6-Tetrahydropyridine, 1,2,5,6-Tetrahydropyridine, 3,6-Dihydro-2H-pyridine, NSC 65443, delta3-Piperidine
IUPAC Name 1,2,3,6-tetrahydropyridine
SMILES C1CC=CCN1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H9N/c1-2-4-6-5-3-1/h1-2,6H,3-5H2
Formula Weight 83.0735
Chemical Name or Material 1,2,3,6-Tetrahydropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.