missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,6-Tetrahydrophthalimide-3,3,4,5,6,6-d6, TRC

Product Code. 30415949
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415949 100 mg 100mg
30417927 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30415949 Supplier Toronto Research Chemicals Supplier No. TRCT294250100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 1,2,3,6-Tetrahydrophthalimide-3,3,4,5,6,6-d6
CAS 1020719-96-3
Purity Grade Notes HPLC
Molecular Formula C8 2H6 H3 N O2
Quantity 100 mg
Synonym 1H-Isoindole-1,3(2H)-dione-4,5,6,7-d4','3a,4,7,7a-tetrahydro-4,7-d2-, 3a,4,7,7a-Tetrahydro-1H-isoindole-1,3(2H)-dione-4,4,5,6,7,7-d6, 1,2,3,6-Tetrahydrophthalimide-d6, 1,2,3,6-Tetrahydrophthalimide-3,3,4,5,6,6-D6, Δ4-Tetrahydrophthalimide-3,3,4,5,6,6-d6, 3a,4,7,7a-Tetrahydro-4,7-d2-1H-isoindole-1,3(2H)-dione-4,5,6,7-d4 (ACI)
InChI Formula InChI=1S/C8H9NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-2,5-6H,3-4H2,(H,9,10,11)/i1D,2D,3D2,4D2
SMILES [2H]C1=C([2H])C([2H])([2H])C2C(C(=O)NC2=O)C1([2H])[2H]
IUPAC Name 4,4,5,6,7,7-hexadeuterio-3a,7a-dihydroisoindole-1,3-dione
Molecular Weight (g/mol) 157.2
Formula Weight 157.101
Percent Purity >95
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.