missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,5-Tetra-O-acetyl beta-D-Ribofuranose, TRC

Product Code. 30179187
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179187 1 g 1g
30179067 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179187 Supplier Toronto Research Chemicals Supplier No. TRCT2831001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13035-61-5
Molecular Formula C13 H18 O9
Molecular Weight (g/mol) 318.28
Synonym β-D-Ribofuranose, 1,2,3,5-tetraacetate, Ribofuranose, tetraacetate, β-D- (8CI), β-D-Ribofuranose, tetraacetate (9CI), 1,2,3,5-Tetra-O-acetyl-β-D-ribofuranose, 1,2,3,5-Tetra-O-acetyl-β-D-ribose, 1,2,3,5-Tetraacetyl-β-D-ribofuranose, 1β-D-Tetraacetylribose, NSC 18738, Tetra-O-acetyl-β-D-ribofuranose, Tetraacetyl-β-D-ribofuranose
IUPAC Name [(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1O[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@@H]1OC(=O)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H18O9/c1-6(14)18-5-10-11(19-7(2)15)12(20-8(3)16)13(22-10)21-9(4)17/h10-13H,5H2,1-4H3/t10-,11-,12-,13-/m1/s1
Formula Weight 318.0951
Chemical Name or Material 1,2,3,5-Tetra-O-acetyl beta-D-Ribofuranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.