missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,5,6,7-Hexahydro-S-5-indacen-4yl-amine, TRC

Product Code. 30393668
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393668 250 mg -
30405334 2.5 g 2.50g
30404773 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393668 Supplier Toronto Research Chemicals Supplier No. TRCH294050250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63089-56-5
Molecular Formula C12 H15 N
Molecular Weight (g/mol) 173.25
IUPAC Name 1,2,3,5,6,7-hexahydro-s-indacen-4-amine
SMILES Nc1c2CCCc2cc3CCCc13

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H15N/c13-12-10-5-1-3-8(10)7-9-4-2-6-11(9)12/h7H,1-6,13H2
Formula Weight 173.1204
Chemical Name or Material 1,2,3,5,6,7-Hexahydro-S-5-indacen-4yl-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.