missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetramethylbenzene, 95%

Product Code. 11399695
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399695 25 g 25g
11389695 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399695 Supplier Thermo Scientific Alfa Aesar Supplier No. L19756.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 488-23-3 | C10H14 | 134.222 g/mol

1,2,3,4-Tetramethylbenzene is used as a starting material in the preparation of 1,2,34-tetramethyl benzene-5-chlorosulfonyl chloride by reacting with chlorosulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetramethylbenzene is used as a starting material in the preparation of 1,2,34-tetramethyl benzene-5-chlorosulfonyl chloride by reacting with chlorosulfonic acid.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-23-3
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008521
InChI Key UOHMMEJUHBCKEE-UHFFFAOYSA-N
Synonym prehnitene, prehnitol, benzene, 1,2,3,4-tetramethyl, tetramethylbenzene, unii-96wt7d2wxj, benzene, tetramethyl, 96wt7d2wxj, ccris 8659, tetramethylbenzenes, tetra-methyl benzene
PubChem CID 10263
ChEBI CHEBI:38997
IUPAC Name 1,2,3,4-tetramethylbenzene
SMILES CC1=C(C(=C(C=C1)C)C)C

Specifications

Density 0.9
Boiling Point 203°C
Flash Point 68°C (154°F)
Refractive Index 1.519
Quantity 25 g
Beilstein 1904390
Solubility Information Immiscible with water.
Formula Weight 134.22
Percent Purity 95%
Chemical Name or Material 1,2,3,4-Tetramethylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.