missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetramethyl-1,3-cyclopentadiene, 85%, mixture of isomers, Thermo Scientific Chemicals

Product Code. p-4510643 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11964431

Brand: Thermo Scientific Acros 348070250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4249-10-9
Molecular Weight (g/mol) 122.21
InChI Key VNPQQEYMXYCAEZ-UHFFFAOYSA-N
Synonym cyanocyclopentane, cyclopentyl cyanide, cyclopentane carbonitrile, cyclopentanenitrile, cyclopentyl nitrile, 1-cyanocyclopentane, cyclopentankohlenitrile, acmc-1aqy6, ksc238k0j, cyclopentanecarbonitrile
PubChem CID 77935
SMILES CC1=C(C)C(C)=C(C)C1

Specifications

Color Yellow
Density 0.8080g/mL
Boiling Point 142.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Packaging Glass bottle
Molecular Formula C9H14
Refractive Index 1.4700 to 1.4760
Quantity 25 g
Specific Gravity 0.808
IUPAC Name cyclopentanecarbonitrile
Formula Weight 122.21
Percent Purity 85%
Physical Form Liquid
Chemical Name or Material 1,2,3,4-Tetramethyl-1,3-cyclopentadiene
Show More Show Less
compliance-icons
Product Identifier
  • 1,2,3,4-Tetramethyl-1,3-cyclopentadiene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.