missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydronaphthalene, Reagent Grade, ≥97%, Honeywell™

Product Code. 15605580 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Stainless Steel Can
Unit Size:
20 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15605580 20 L Stainless Steel Can 20 L
15638410 20 L Stainless Steel Can 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15605580 Supplier Honeywell Research Chemicals Supplier No. 45643820L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 119-64-2
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00001733
InChI Key CXWXQJXEFPUFDZ-UHFFFAOYSA-N
Synonym tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene
PubChem CID 8404
ChEBI CHEBI:35008
IUPAC Name 1,2,3,4-tetrahydronaphthalene
SMILES C1CCC2=CC=CC=C2C1

Specifications

Melting Point -35°C
Color Colorless
Density 0.973g/mL at 25°C
Boiling Point 207°C
Flash Point 71°C (159.8°F)
Infrared Spectrum Conforms to structure
Packaging Stainless Steel Can
Refractive Index n20/D 1.541
Quantity 20 L
UN Number UN3082
Beilstein 1446407
Formula Weight 132.2g/mol
Percent Purity ≥97%
Chemical Name or Material 1,2,3,4-Tetrahydronaphthalene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.