missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydronaphthalene, 98+%, ACROS Organics™

Brand:  Acros Organics 146730100

Code : 29

Additional Details : CAS Number : 119-64-2 Weight : 9.78000kg Transport : UN number : 3082 Chem class : 1 Pack group : III

Quantity 10L
Packaging Metal Drum

 View more versions of this product

Product Code. 10423255

  • £268.60 / 10 L


Estimated Shipment
20-02-2018
Add to basket

Specifications

Chemical Identifiers

1,2,3,4-Tetrahydronaphthalene
98.0
Pure
C10H12
132.2
THN
119-64-2
100.0
98+%
C10H12
MFCD00001733

Specifications

1,2,3,4-Tetrahydronaphthalene
77°C
207.0°C
10L
98.0
Metal Drum
Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents
98+%
C10H12
132.2
05, IV, 1388
15, 9368
0.9730g/mL
0.973
− 35.0°C
119-64-2
100.0
1.5400 to 1.5420
Pure
98+%
C10H12
MFCD00001733
01,1142
THN
Videos
SDS