missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroisoquinoline-1-acetic acid, 98%, Thermo Scientific™

Product Code. 10206111 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10206111 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10206111 Supplier Thermo Scientific Acros Supplier No. 334380010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 105400-81-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 248.0°C to 251.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C11H13NO2
MDL Number MFCD00215024
Quantity 1 g
Synonym 1,2,3,4-tetrahydroisoquinoline-1-yl acetic acid, 2-1,2,3,4-tetrahydroisoquinolin-1-yl acetic acid, 1,2,3,4-tetrahydroisoquinoline-1-acetic acid, 1,2,3,4-tetrahydro-isoquinolin-1-yl-acetic acid, 2-1,2,3,4-tetrahydro-1-isoquinolinyl acetic acid, 1,2,3,4-tetrahydroisoquinolin-1-ylacetic acid, 1-isoquinolineaceticacid, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro-1-isoquinolinylacetic acid, 2-1,2,3,4-tetrahydroisoquinolyl acetic acid, tetrahydroisoquinolylacetic acid
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol
InChI Key BUAVPRGEIAVFBF-JTQLQIEISA-N
SMILES [O-]C(=O)C[C@@H]1[NH2+]CCC2=CC=CC=C12
IUPAC Name 2-(1,2,3,4-tetrahydroisoquinolin-1-yl)acetic acid
Molecular Weight (g/mol) 191.23
PubChem CID 2733683
Formula Weight 191.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,4-Tetrahydroisoquinoline-1-acetic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.