missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydrobenzazepine-5-one, 97%, Thermo Scientific™

Product Code. 10372110 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10372110

Brand: Thermo Scientific Acros 435910050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1127-74-8
MDL Number MFCD03426404
InChI Key NKRKBYFBKLDCFB-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-benzo b azepin-5-one, 1,2,3,4-tetrahydrobenzo b azepin-5-one, 1,2,3,4-tetrahydrobenzoazepine-5-one, 3,4-dihydro-1h-benzo b azepin-5 2h-one, 1,2,3,4-tetrahydro-5h-1-benzazepin-5-one, 2,3,4,5-tetrahydro-1h-1-benzazepin-5-one, 5h-1-benzazepin-5-one, 1,2,3,4-tetrahydro, 5-oxo-2,3,4,5-tetrahydro-1h-benzazepine, 1,2,3,4-tetrahydrobenzazepine-5-one, zlchem 1002
PubChem CID 294788
IUPAC Name 1,2,3,4-tetrahydro-1-benzazepin-5-one
SMILES C1CC(=O)C2=CC=CC=C2NC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 68°C to 72°C
Color Green to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C10H11NO
Quantity 5 g
Molecular Weight (g/mol) 161.204
Formula Weight 161.21
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 1,2,3,4-Tetrahydrobenzazepine-5-one
Show More Show Less
compliance-icons
Product Identifier
  • 1,2,3,4-Tetrahydrobenzazepine-5-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.