missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-1-naphthoic acid, 98%

Product Code. 15454827
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454827 250 mg 250mg
15444827 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454827 Supplier Thermo Scientific Alfa Aesar Supplier No. H32138.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1914-65-4 | C11H12O2 | 176.215 g/mol

1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.

Solubility
Soluble in methanol, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1914-65-4
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD02179127
InChI Key VDLWTJCSPSUGOA-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydro, tetralin-1-carboxylic acid, 1,2,3,4-tetrahedro-naphthoic acid, 1,2,3,4-tetrahydro-naphthalene-1-carboxylic acid, chembl81606, 1-tetralincarboxylic acid, 1,2,3,4-tetrahydro-1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydronaphthalenecarboxylic acid, tetrahydronaphthoic acid
PubChem CID 12598146
IUPAC Name 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
SMILES C1CC(C2=CC=CC=C2C1)C(=O)O

Specifications

Melting Point 82°C to 85°C
Quantity 250 mg
Solubility Information Soluble in methanol,and chloroform.
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material 1,2,3,4-Tetrahydro-1-naphthoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.