missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-1-naphthalenecarboxylic Acid, Mikromol™

Product Code. 30051146
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30051146 100 mg 100mg
30049882 25 mg 25mg
2 options
This item is not returnable. View return policy

Product Code. 30051146

Brand: Mikromol MM0088.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,2,3,4-Tetrahydro-1-naphthalenecarboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,3,4-Tetrahydro-1-naphthalenecarboxylic Acid
CAS 1914-65-4
Analyte or Component Names 1,2,3,4-Tetrahydro-1-naphthalenecarboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C11 H12 O2
Certifications/Compliance ISO 17025
Synonym 1,2,3,4-Tetrahydro-1-naphthalenecarboxylic Acid
InChI Formula InChI=1S/C11H12O2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6,10H,3,5,7H2,(H,12,13)
SMILES OC(=O)C1CCCc2ccccc12
IUPAC Name 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Molecular Weight (g/mol) 176.21
Quantity 100 mg
Formula Weight 176.0837 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.