missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Butanetetracarboxylic acid, 99+%

Product Code. 10350484 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10350484 500 g Glass bottle 500g
10602423 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10350484 Supplier Thermo Scientific Acros Supplier No. 366765000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1703-58-8 | C8H10O8 | 234.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1703-58-8
Molecular Formula C8H10O8
Molecular Weight (g/mol) 234.16
MDL Number MFCD00002722
InChI Key GGAUUQHSCNMCAU-UHFFFAOYSA-N
Synonym 1,2,3,4-butanetetracarboxylic acid, butanetetracarboxylic acid, dsstox_cid_4670, dsstox_rid_77494, dsstox_gsid_24670, 1,2,3,4-butanetetracarboxylic, meso-1,2,3,4-butanetetracarboxylic acid, acmc-20abw3, ksc176c1b, ggauuqhscnmcau-uhfffaoysa
PubChem CID 15560
IUPAC Name butane-1,2,3,4-tetracarboxylic acid
SMILES C(C(C(CC(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 195.0°C to 197.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 500 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 234.16
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Butanetetracarboxylic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2,3,4-Butanetetracarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.