missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, TRC

Product Code. 30413553
Change view
Click to view available options
Quantity:
25 g
250 g
Unit Size:
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413553 25 g -
30421670 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30413553 Supplier Toronto Research Chemicals Supplier No. TRCP23010025G
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25941-03-1
Molecular Formula C16 H22 O11
Molecular Weight (g/mol) 390.34
IUPAC Name [(2R,3R,4S,5S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1OC(OC(=O)C)[C@@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C16H22O11/c1-7(17)22-6-12-13(23-8(2)18)14(24-9(3)19)15(25-10(4)20)16(27-12)26-11(5)21/h12-16H,6H2,1-5H3/t12-,13-,14+,15+,16?/m1/s1
Formula Weight 390.1162
Chemical Name or Material 1,2,3,4,6-Penta-O-acetyl-D-mannopyranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.