missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,6,7,8-Heptachlorodibenzofuran, TRC

Product Code. 30395540
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395540 1 mg -
30405502 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395540 Supplier Toronto Research Chemicals Supplier No. TRCH2650801MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67562-39-4
Molecular Formula C12 H Cl7 O
Molecular Weight (g/mol) 409.31
Synonym Dibenzofuran, 1,2,3,4,6,7,8-heptachloro-, 1,2,3,4,6,7,8-Heptachlorodibenzofuran, 1,2,3,4,6,7,8-HpCDF, 1234678-HpCDF, F 131, HpCDF, PCDF 131, 1,2,3,4,6,7,8-HeptaCDF, 1234678-HeptaCDF, 1234678-HpCDF, 1234678-Heptachlorodibenzofuran
IUPAC Name 1,2,3,4,6,7,8-heptachlorodibenzofuran
SMILES Clc1cc2c(oc3c(Cl)c(Cl)c(Cl)c(Cl)c23)c(Cl)c1Cl

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C12HCl7O/c13-3-1-2-4-6(15)7(16)8(17)10(19)12(4)20-11(2)9(18)5(3)14/h1H
Formula Weight 405.7847
Chemical Name or Material 1,2,3,4,6,7,8-Heptachlorodibenzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.