missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5,6,7-Heptachloronaphthalene (>90%), TRC

Product Code. 30389578
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389578 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389578 Supplier Toronto Research Chemicals Supplier No. TRCH26521010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,3,4,5,6,7-Heptachloronaphthalene
CAS 58863-14-2
Purity Grade Notes HPLC
Molecular Formula C10 H Cl7
Quantity 10 mg
Synonym 1H-Heptachloronaphthalene, PCN 73
InChI Formula InChI=1S/C10HCl7/c11-3-1-2-4(7(14)6(3)13)8(15)10(17)9(16)5(2)12/h1H
SMILES Clc1cc2c(Cl)c(Cl)c(Cl)c(Cl)c2c(Cl)c1Cl
IUPAC Name 1,2,3,4,5,6,7-heptachloronaphthalene
Molecular Weight (g/mol) 369.29
Formula Weight 365.7898
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.