missing translation for 'onlineSavingsMsg'
Learn More

1,2,2,6,6-Pentamethylpiperidine, 95%

Product Code. 10493741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10493741 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10493741 Supplier Thermo Scientific Acros Supplier No. 416960050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-55-0 | C10H21N | 155.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 79-55-0
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8580g/mL
Boiling Point 187.0°C to 188.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C10H21N
Refractive Index 1.4570 to 1.4600
MDL Number MFCD00006493
Quantity 5 g
Merck Index 15, 7189
Specific Gravity 0.858
Synonym pempidine, pyrilene, perolysen, tenormal, piperidine, 1,2,2,6,6-pentamethyl, pempidina dcit, pempidine inn:ban, pempidinum inn-latin, unii-n5i18ji9d6, 1,2,2,6,6-pentamethyl-piperidine
InChI Key XULIXFLCVXWHRF-UHFFFAOYSA-N
SMILES CC1(CCCC(N1C)(C)C)C
IUPAC Name 1,2,2,6,6-pentamethylpiperidine
Molecular Weight (g/mol) 155.28
PubChem CID 6603
Formula Weight 155.28
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1,2,2,6,6-Pentamethylpiperidine, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2,2,6,6-Pentamethylpiperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.